Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SENP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | SENP3 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15659020
![]() |
Novus Biologicals
NBP15659020UL |
20 μL |
Each for $158.00
|
|
|||||
NBP156590
![]() |
Novus Biologicals
NBP156590 |
100 μL |
Each for $487.50
|
|
|||||
Description
SENP3 Polyclonal specifically detects SENP3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
SENP3 | |
Unconjugated | |
RUO | |
26168 | |
Synthetic peptides corresponding to SENP3(SUMO1/sentrin/SMT3 specific peptidase 3) The peptide sequence was selected from the N terminal of SENP3. Peptide sequence PPPKPRLKSGGGFGPDPGSGTTVPARRLPVPRPSFDASASEEEEEEEEEE. | |
Primary |
Polyclonal | |
Rabbit | |
DKFZP586K0919, DKFZp762A152, EC 3.4.22, EC 3.4.22.-, sentrin/SUMO-specific protease 3, Sentrin/SUMO-specific protease SENP3, sentrin-specific protease 3, SMT3IP1, SSP3DKFZp586K0919, SUMO1/sentrin/SMT3 specific peptidase 3, SUMO1/sentrin/SMT3 specific protease 3, SUMO-1-specific protease 3, SUSP3 | |
SENP3 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title