Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SENP5 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056412 0.1 mg
NB10056412T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056412 Supplier Novus Biologicals Supplier No. NB10056412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SENP5 Polyclonal specifically detects SENP5 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen SENP5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q96HI0
Gene Alias DKFZp564O1016, EC 3.4.22, EC 3.4.22.-, FLJ42398, MGC27076, Sentrin/SUMO-specific protease SENP5, sentrin-specific protease 5, SUMO1/sentrin specific peptidase 5, SUMO1/sentrin specific protease 5
Gene Symbols SENP5
Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 229-246 (CNRKPFINREITNYRARH) of human SENP5
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 205564
Test Specificity The antibody should recognize SENP5 isoforms containing the immunogen sequence (CNRKPFINREITNYRARH) used to make the antibody. Isoform sizes from 163-755 amino acids have been described for SENP5. Researchers are encouraged to BLAST the sequence (NCBI.NIH.GOV) to gain additional information.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.