Learn More
Sensidyne™ Accessories for Sampling Media
$105.30 - $1229.65
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19-001-516
![]()
|
Sensidyne™
800232 |
Tube Holder Deluxe Kit |
Each for $1,229.65
|
|
|||||
19-001-517
![]()
|
Sensidyne™
800252 |
Tube Holder Starter Kit |
Each for $493.65
|
|
|||||
19-031427
![]()
|
Sensidyne™
800143 |
Filter Cassette Holder |
Each for $105.30
|
|
|||||
Description
Sampling media accessories include filter cassette holder that attaches to a worker's collar to sample breathing zone.
Tube holder starter kit includes single-variable manifold, holders, ends, and tubing. Tube holder deluxe kit includes single-, dual-, and triple-variable manifolds; holders; ends; and tubing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.