Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sensidyne™ Detector Tubes: Carbon Dioxide
SDP

Catalog No. p-177798 Shop All Sensidyne Inc. Products
Encompass
Click to view available options
Measurement Range:
0.01 to 0.70%
0.1 to 5.2%
100 to 4000
5 to 50%
Response Time:
1.5 min.
2 min.
5 min.
Not Applicable
No. of Strokes:
1/2
n/a

Catalog No. 18-067W

Supplier: Sensidyne™ 126UH

Only null left
Add to Cart
Add to Cart

Safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.

  • Easy to use detector tubes
  • Provides low-cost method of routine or spot sampling for gases and vapors
  • Sold in packs of 10

Specifications

Components Measured Carbon Dioxide
Measurement Range 5 to 50%
No. of Strokes 1/2
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.