Learn More
Sensidyne™ Detector Tubes: Acetone
Detector tubes
Supplier: Sensidyne™ 102SC
Description
All detector tubes are sold in packs of 10.Specifications
Acetone | |
1 | |
1 min. | |
0°C to 40°C |
0.01 to 4% | |
10 | |
32°F to 104°F |