Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sensidyne™ Detector Tubes: Ammonia
SDP

Catalog No. p-6474180 Shop All Sensidyne Inc. Products
Encompass
Change view
Click to view available options
Measurement Range:
0.1 to 1%
0.2 to 20
5 to 260
Response Time:
0.5 min.
1 min.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time
19-818-940 0.1 to 1% 0.5 min.
18-067A 0.2 to 20 1 min.
18-067B 5 to 260 1 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-818-940 Supplier Sensidyne™ Supplier No. 105SM
Only null left
Add to Cart
Add to Cart

Safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.

  • Easy to use detector tubes
  • Provides low-cost method of routine or spot sampling for gases and vapors
  • Sold in packs of 10

Specifications

Components Measured Ammonia
Measurement Range 0.1 to 1%
No. of Strokes 1
Quantity 10
Response Time 0.5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.