Learn More
Sensidyne™ Gilian™ LFS-113 Air Sampling System
SDP
Includes: Rechargeable NiCd battery pack (included) provides eight hours of continuous operation.
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-087531
SDP
View Documents
Sensidyne™
800088111
Without Clock Without Clock N/A N/A N/A
19-087532
SDP
View Documents
Sensidyne™
800098111
With Clock With Clock N/A N/A N/A
Description
Description
Dual-mode, low-flow air sampler
Personal low-flow air samplers ideal for sorbent tube collection of gases and vapors (18-556V and 18-556W sorbent tubes sold separately). Feature two modes of operation: constant- flow mode (5 to 200cc/min.) for single-tube sampling and constant-pressure mode (1 to 350cc/min.) for multiple-tube sampling.
Instant fault function automatically stops internal pump when sampler is operated outside its performance envelope. EMI/RFI shielding case protects instrument from electromagnetic and radio-frequency interference.
Order Info
Compact, lightweight unit features belt clip. Sampler measures approximately 2 1/2L x 1 1/2W x 4 1/2 in.H (6.4 x 3.6 x 11.7cm). Net weight: 12 oz. (340g). Unit is available with or without a time-elapsed clock.
Certifications
UL listed, UL listed as intrinsically safe for use in Class I, Groups A, B, C, and D; Class II, Groups E, F, and G; and Class III hazardous locations.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.