Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Separase Antibody (XJ11-1B12), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB100439G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100439G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100439G Supplier Novus Biologicals Supplier No. NB100439G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Separase Monoclonal specifically detects Separase in Human samples. It is validated for Western Blot.

Specifications

Antigen Separase
Applications Western Blot
Classification Monoclonal
Clone XJ11-1B12
Conjugate DyLight 488
Dilution Western Blot
Gene Accession No. Q14674
Gene Alias Caspase-like protein ESPL1, EC 3.4.22.49, ESP1extra spindle poles like 1, extra spindle pole bodies homolog 1 (S. cerevisiae), extra spindle poles like 1 (S. cerevisiae), Extra spindle poles-like 1 protein, FLJ46492, KIAA0165separin, Separase
Gene Symbols ESPL1
Host Species Mouse
Immunogen Maltose-Binding Protein fusion of a C-terminal fragment of human Separase (residues 1866-1996). [UniProt# Q14674]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9700
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.