Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Sephin 1

Click to view available options
Quantity:
10 mg
50 mg
Description
Sephin 1 is an inhibitor of the regulatory subunit PPP1R15A of protein phosphatase 1. Does not bind PPP1R15B or have α2 adrenergic agonist activity. Cytoprotective in endoplasmic reticulum stressed cells in vitro. Prevents defects associated with protein misfolding in a mouse Charcot-Marie Tooth-1B model. Orally available.
Specifications
Specifications
| Chemical Name or Material | (2E)-2-[(2-Chlorophenyl)methylene]-hydrazinecarboximidamide carbonic acid salt |
| Quantity | 10 mg |
| Target | Protein Ser/Thr Phosphatase Inhibitors |
| Molecular Formula | C8H9ClN4.CH2O3 |
| Formula Weight | 258.66 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction