Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sephin 1
GREENER_CHOICE

Catalog No. p-7110845
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of the regulatory subunit PPP1R15A of protein phosphatase 1

Sephin 1 is an inhibitor of the regulatory subunit PPP1R15A of protein phosphatase 1. Does not bind PPP1R15B or have α2 adrenergic agonist activity. Cytoprotective in endoplasmic reticulum stressed cells in vitro. Prevents defects associated with protein misfolding in a mouse Charcot-Marie Tooth-1B model. Orally available.

Specifications

Chemical Name or Material (2E)-2-[(2-Chlorophenyl)methylene]-hydrazinecarboximidamide carbonic acid salt
Quantity 10 mg
Target Protein Ser/Thr Phosphatase Inhibitors
Molecular Formula C8H9ClN4.CH2O3
Formula Weight 258.66
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.