Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEPN1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP169058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP169058 100 μL
NBP16905820 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP169058 Supplier Novus Biologicals Supplier No. NBP169058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SEPN1 Polyclonal specifically detects SEPN1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SEPN1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. D3Z5N3
Gene Alias FLJ24021, rigid spine muscular dystrophy 1, RSMD1, RSS, selenoprotein N, selenoprotein N, 1, SelN, SELNMDRS1
Gene Symbols SEPN1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to Sepn1 (selenoprotein N, 1) The peptide sequence was selected from the C terminal of Sepn1. Peptide sequence VHHINANYFLDITSMKPEDMENNNVFSFSSSFEDPSTATYMQFLREGLRR.
Molecular Weight of Antigen 62 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57190
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Equine, Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.