Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-1 Antibody (OTI1E1), HRP, Novus Biologicals™
SDP

Catalog No. NBP274072H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274072H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274072H Supplier Novus Biologicals Supplier No. NBP274072H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Septin-1 Monoclonal specifically detects Septin-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Septin-1
Applications Western Blot
Classification Monoclonal
Clone OTI1E1
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias DIFF6SEP1, differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase), LARP, Peanut-like protein 3, PNUTL3MGC20394, septin 1, septin-1, Serologically defined breast cancer antigen NY-BR-24
Gene Symbols SEPT1
Host Species Mouse
Immunogen Full length recombinant protein of human Septin-1 (NP_443070) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 1731.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.