Learn More
Description
Specifically detects Septin-10 in Human samples, and it is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | Septin-10 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate |
| Formulation | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide |
| Gene Alias | FLJ11619, septin 10, septin-10 |
| Gene Symbols | 43353 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
