Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-10 Antibody, Novus Biologicals™
SDP

Catalog No. NBP244288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244288 Supplier Novus Biologicals Supplier No. NBP244288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Septin-10 in Human samples, and it is validated for Immunoprecipitation

Septin-10 Polyclonal specifically detects Septin-10 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen Septin-10
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias FLJ11619, septin 10, septin-10
Gene Symbols 43353
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residue 404 to 454 of human Septin-10 using the numbering given in entry Q9P0V9.2 (GeneID 151011).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 151011
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.