Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-12 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP15811720UL

 View more versions of this product

Catalog No. NBP1581720


Only null left
Add to Cart

Description

Description

Septin-12 Polyclonal specifically detects Septin-12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Septin-12
Polyclonal
Western Blot 1:100-1:2000
Q8IYM1
43355
Synthetic peptides corresponding to Septin-12. The peptide sequence was selected from the middle region of 40433. Peptide sequence LQRLCRTVNVVPVIARADSLTMEEREAFRRRIQQNLRTHCIDVYPQMCFD.
20 μL
Cell Cycle and Replication
124404
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
FLJ25410, septin 12, septin-12
Rabbit
Affinity Purified
RUO
Primary
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.