Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-2 Rabbit anti-Human, Mouse, Rat, Clone: JB41-30, Novus Biologicals™
SDP

Catalog No. NBP275660 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275660 Supplier Novus Biologicals Supplier No. NBP275660

Rabbit Monoclonal Antibody

Septin-2 Monoclonal specifically detects Septin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Septin-2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JB41-30
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DIFF6NEDD-5, hNedd5, KIAA0158Neural precursor cell expressed developmentally down-regulated protein 5, NEDD5Pnutl3, neural precursor cell expressed, developmentally down-regulated 5, septin 2, septin-2
Host Species Rabbit
Immunogen Recombinant protein within Human Septin-2 aa 182-318/ 361. (SwissProt: Q15019 Human; SwissProt: P42208 Mouse; SwissProt: Q91Y81 Rat)
Molecular Weight of Antigen 41 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4735.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.