Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357643 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357643 Supplier Novus Biologicals Supplier No. NBP176667MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Septin-4 Polyclonal antibody specifically detects Septin-4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Septin-4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Apoptosis-related protein in the TGF-beta signaling pathway, ARTSPeanut-like protein 2, BRADEION, Bradeion beta, Brain protein H5, CE5B3, Cell division control-related protein 2, Cerebral protein 7, H5, hCDCREL-2SEP4, hucep-7, MART, peanut-like 2, peanut-like 2 (Drosophila), PNUTL2CE5B3 beta, septin 4, septin-4, septin-M
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to amino acids near the amino terminus of human ARTS. The immunogen is located within the first 50 amino acids of ARTS. Amino Acid Squence: KRFLEDTTDDGE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 5414
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.