Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152096 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152096 Supplier Novus Biologicals Supplier No. NBP152096
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Septin-4 Polyclonal specifically detects Septin-4 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Septin-4
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1-0.3 ug/mL, Flow Cytometry 10 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004565.1
Gene Alias Apoptosis-related protein in the TGF-beta signaling pathway, ARTSPeanut-like protein 2, BRADEION, Bradeion beta, Brain protein H5, CE5B3, Cell division control-related protein 2, Cerebral protein 7, H5, hCDCREL-2SEP4, hucep-7, MART, peanut-like 2, peanut-like 2 (Drosophila), PNUTL2CE5B3 beta, septin 4, septin-4, septin-M
Gene Symbols 43347
Host Species Goat
Immunogen Peptide with sequence C-ELSKFVKDFSGN corresponding to internal region according to NP_004565.1, NP_536340.1, NP_536341.1, NP_001185642.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 5414
Test Specificity This antibody is expected to recognize all reported isoforms (NP_004565.1; NP_536340.1; NP_536341.1, NP_001185642.1). Amino acid numbering in name refers to NP_004565.1 sequence.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.