Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-5 Antibody (SP18), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB015026 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015026 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015026 Supplier Novus Biologicals Supplier No. NBP281056AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Septin-5 Monoclonal specifically detects Septin-5 in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Septin-5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone SP18
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5
Gene Symbols SEPTIN5
Host Species Rabbit
Immunogen Immunoprecipitate from human brain.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5413
Test Specificity This antibody recognises Septin-5 (CDCrel-1) and does not cross-react with CDCrel-2.
Target Species Human, Rat, Rabbit
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.