Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-5 Antibody (SP18), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342927 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342927 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342927 Supplier Novus Biologicals Supplier No. NBP281058MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Septin-5 Monoclonal antibody specifically detects Septin-5 in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen Septin-5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone SP18
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5
Host Species Mouse
Immunogen Immunoprecipitate from human brain.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5413
Target Species Human, Rat, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.