Learn More
Septin 9 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | Septin 9 |
---|---|
Applications | Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2512
![]() |
Bethyl Laboratories, Inc
IHC00672T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2511
![]() |
Bethyl Laboratories, Inc
IHC00672 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Monkey, Crab-eating macaque and White-tufted-ear marmoset
The maf oncogene was identified by structural analysis of the AS42 avian transforming retrovirus genome. The Maf family is divided into two subclasses, large Mafs (vMaf, cMaf, MafB and Nrl) and small Mafs (MafF, MafK, and MafG). Both subclasses contain leucinezipper motifs, which allow homodimerization as well as heterodimerization with a variety of other bZip transcription factors. Large Mafs also contain an acidic transactivation domain absent in the small Maf proteins. Although they do not possess inherent transactivation activity, small Maf proteins can act as positive regulators of transcription by targeting transcriptionally active dimerization partners to specific DNA regulatory elements. Conversely, small Mafs can act also as negative regulators of transcription by recruiting transcriptional repressors or by forming homodimers that can replace active dimers. Human MafF was isolated in a yeast one-hybrid system from a human myometrium cDNA library. Human MAFF encodes a 164 amino acids proten. Like other small MAFF proteins, it contains an extended leucine zipper structure and lacks an N-terminal transactivating domain. The three small Maf proteins have been implicated in a number of physiological processes, including development, differentiation, haematopoiesis and stress response. Interestingly, these three proteins regulate the stress response via different mechanisms.Specifications
Septin 9 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
AF17q25, KIAA0991, MLL septin-like fusion, MLL septin-like fusion protein, MLL septin-like fusion protein MSF-A, MSF, MSF1, NAPB, Ov/Br septin, ovarian/breast septin, PNUTL4, SeptD1, septin D1, SINT1 | |
SEPT9 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9UHD8 | |
10801 | |
Between 536 and 586 | |
Primary | |
4° C | |
SEPT9 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.