Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ SeqStudio™ 24 Flex Genetic Analyzer
GSA_VA

Catalog No. p-200019980
Encompass_Preferred
Click to view available options
External Computer:
Not included

The Applied Biosystems SeqStudio 24 Flex Genetic Analyzer offers design improvements and technological advances to the capillary electrophoresis (CE) workflow.

The Applied Biosystems SeqStudio 24 Flex Genetic Analyzer offers design improvements and technological advances to the capillary electrophoresis (CE) workflow. It delivers gold-standard Sanger sequencing and fragment analysis data quality with increased flexibility, easier operation, and cloud connectivity to enable researchers to manage their workloads and work/life balance effectively. Designed for ultimate ease-of-use, this mid-throughput system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.

Features of the SeqStudio 24 Flex Genetic Analyzer include:
• Space savings with true standalone operation; setup, start, monitor, and analyze directly on the touchscreen
• Improve workload management with continuous plate loading and sample reprioritization
• Realize lab efficiencies with remote monitoring
• New, improved capillary arrays for simple and hassle-free handling
• Reduce downtime with integrated remote troubleshooting tools

Easy-to-use data collection software
The data collection software pre-installed on the on-board touch screen computer takes user-friendliness to a new level. It offers an intuitive dashboard design, simplified plate set-up, and embedded self-help videos for common operations so that new users can get up and running quickly.

Scheduling flexibility
Four-plate capacity, continuous plate loading, and urgent sample reprioritization provide both the largest mid-throughput capacity and complete scheduling flexibility, translating into extra time in your day.

Remote functionality
Our cloud-based Connect platform provides enhanced flexibility by allowing users to set up plate files, monitor instrument and consumables performance and run progress, and even perform some critical instrument functions from anywhere.

Stay connected to your data
The SeqStudio 24 Flex system offers an automated workflow to obtain and share results from existing secondary analysis applications on Connect. These web-based software applications include Peak Scanner, Microsatellite Analysis, NGS Confirmation, Quality Check and Variant Analysis. They are compatible with Mac and PC.

Peace of mind with Smart service
Minimize downtime with premium service offerings for the SeqStudio 24 Flex Genetic Analyzer. Smart Help allows users to contact technical support directly from the touchscreen, without the need to download files or spend time on the phone. Experienced technicians will analyze the request and respond according to the user’s preference (phone or email) and resolve the issue as quickly as possible.

Smart Remote Support is an immersive collaboration tool that allows customers to show issues quickly and efficiently to technical support for resolution, helping reduce instrument downtime and minimize the need for on-site visits.

High application versatility
Simple workflows, reliable assays and reagents, and intuitive software are available for a broad range of applications, including multiplexed PCR analysis, next-generation confirmation, species identification, STR genotyping, cell-line authentication, dsDNA quality control, plasmid sequencing, and genome editing analysis.

Results you can trust
Well-to-well and instrument-to-instrument data accuracy is achieved with the proven Applied Biosystems capillary electrophoresis technology.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description SeqStudio™ 24 Flex
Capillary Length 36 or 50 cm
For Use With (Equipment) Thermal Cyclers
External Computer Not included
Filtered Filtered
Format 96-well plates, 384-well plates, or 8-tube strips
High-throughput Compatibility No
Operating Environment 59–86°F (15–30°C), 20–80% relative humidity (non-condensing). Room temperature should not fluctuate ±2°C during an instrument run.
Polymer POP-4™, POP-6™, POP-7™
Power Requirements 100–240 ± 10% VAC; 50/60 Hz
Quantity Each
Shipping Condition Environmentally controlled
Sterility Non-sterile
Temperature Range (Metric) 18–70°C (Oven temperature control)
Dimensions 27.6 x 26.6 x 34.1 in. (70 x 67.5 x 86.5 cm)
Weight 253.5 lb. (115 kg)
Voltage 100/240 V
Total Service Coverage Standard 1-year warranty included, extended warranties available
Max. Power Electrophoresis Voltage Up to 20 kV
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.