Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SeQuant™ ZIC™-HILIC Analytical PEEK Columns
GSA_VA

Catalog No. M1504520001 Shop All MilliporeSigma Products
Change view
Click to view available options
Length (Metric):
20 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
Particle Size:
3.5 μm
5 μm
Porosity:
100 Å
200 Å
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Particle Size Porosity
M1504520001 100 mm 2.1 mm 5 μm 200 Å
M1504440001 150 mm 4.6 mm 3.5 μm 100 Å
M1504420001 150 mm 2.1 mm 3.5 μm 100 Å
M1504500001 50 mm 4.6 mm 5 μm 200 Å
M1504540001 150 mm 2.1 mm 5 μm 200 Å
M1504480001 150 mm 2.1 mm 3.5 μm 200 Å
M1504530001 100 mm 4.6 mm 5 μm 200 Å
M1504400001 50 mm 2.1 mm 3.5 μm 100 Å
M1504550001 150 mm 4.6 mm 5 μm 200 Å
M1504490001 150 mm 4.6 mm 3.5 μm 200 Å
M1504430001 250 mm 2.1 mm 3.5 μm 100 Å
M1504570001 250 mm 2.1 mm 5 μm 200 Å
M1504460001 50 mm 4.6 mm 3.5 μm 200 Å
M1504470001 100 mm 2.1 mm 3.5 μm 200 Å
M1504450001 50 mm 2.1 mm 3.5 μm 200 Å
M1504580001 250 mm 4.6 mm 5 μm 200 Å
M1504390001 20 mm 2.1 mm 3.5 μm 100 Å
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. M1504520001 Supplier MilliporeSigma™ Supplier No. 1504520001
Add to Cart
Add to Cart

PEEK coated HPLC column

SeQuant™ ZIC™-HILIC is the ideal choice for separation of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase, ensures reproductible retention of all types of compounds that have poor retention and are difficult to separate on reversed-phase HPLC columns. ZIC™-HILIC is designed for compounds such as acids and bases, anions and cations, carbohydrates, metabolites, metal complexes, amino acids, peptides, protein digests and much more

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 100 mm
Particle Size 5 μm
Pore Size 200 Å
Porosity 200 Å
Quantity 1/Pk.
Material Metal-free PEEK
Product Type Column
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.