Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Sera-Mag™ SpeedBeads Streptavidin-Blocked Magnetic Particles
Shop All Cytiva Products
Click to view available options
Volume (Metric):
1 mL
10 mL
Description
- Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles combine high affinity for biotinylated target molecules with very low nonspecific binding.
- Very high binding capacity and low non-specific binding for biotinylated target molecules on-surfactant due to non-protein blocking reagent
- Fast reaction kinetics increases throughput and precision, enabling faster movement through viscous solutions
- Unique cauliflower-like surface structure provides increased surface area for binding reactions compared to smooth-surfaced particles
- Uniform, nominal 1 μm and 3 μm diameters provide excellent lot-to-lot reproducibility
- Also available as custom manufactured beads
Specifications
Specifications
| Concentration | 10 mg/mL |
| Content And Storage | 4°C |
| Contents | Magnetic Beads |
| Color | Black |
| Material | Magnetite |
| Product Type | Dye Reagents |
| Volume (Metric) | 10 mL |
| Diameter (Metric) | 3 μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction