Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads
Shop All Cytiva Products
Click to view available options
Quantity:
5 mL
60 mL
Size:
400 nm diameter particle
400 nm diameter particles
700 nm diameter particle
Description
Specifications
Specifications
| Concentration | 20 mg/mL in water (0.05% sodium azide) |
| Content And Storage | Room Temperature |
| Description | Storage Solution: Water + 0.05% sodium azide |
| Diameter (Metric) | 400 nm |
| For Use With (Application) | Chaotropic salt chemistry and vortex mixing, roller mixing and sonication |
| Product Type | Silica coated, core-shell structure |
| Quantity | 5 mL |
| Size | 400 nm diameter particles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction