Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERBP1 Antibody (OTI5G2), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP274073A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274073A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274073A94 Supplier Novus Biologicals Supplier No. NBP274073AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERBP1 Monoclonal specifically detects SERBP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SERBP1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5G2
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CGI-55, CHD3IP, chromodomain helicase DNA binding protein 3 interacting protein, DKFZP564M2423, HABP4L, PAI-1 mRNA binding protein, PAI1 RNA-binding protein 1, PAI-RBP1DKFZp564M2423, PAIRBP1FLJ90489, plasminogen activator inhibitor 1 RNA binding protein, plasminogen activator inhibitor 1 RNA-binding protein, SERPINE1 mRNA binding protein 1, SERPINE1 mRNA-binding protein 1
Gene Symbols SERBP1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-262 of human SERBP1 (NP_001018078) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 26135
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.