Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERCA2 ATPase Antibody (IID8), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB30529A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30529A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30529A350 Supplier Novus Biologicals Supplier No. NB300529AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SERCA2 ATPase Monoclonal specifically detects SERCA2 ATPase in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen SERCA2 ATPase
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IID8
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2
Gene Symbols ATP2A2
Host Species Mouse
Immunogen Purified canine cardiac sarcoplasmic reticulum.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 488
Test Specificity Detects Sarcoplasmic or Endoplasmic Reticulum Calcium 2 (SERCA2) ATPase. This does not discriminate between the two isoforms.
Target Species Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.