Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERCA2 ATPase Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB045209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045209 Supplier Novus Biologicals Supplier No. NBP297582JF549

Rabbit Polyclonal Antibody

SERCA2 ATPase Polyclonal specifically detects SERCA2 ATPase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SERCA2 ATPase
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SERCA2 ATPase fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 488
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.