Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERCA3 ATPase Antibody, Novus Biologicals™
SDP

Catalog No. NBP220306 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220306 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220306 Supplier Novus Biologicals Supplier No. NBP220306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERCA3 ATPase Polyclonal specifically detects SERCA3 ATPase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SERCA3 ATPase
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Accession No. Q93084
Gene Alias adenosine triphosphatase, calcium, ATPase, Ca(2+)-transporting, ubiquitous, ATPase, Ca++ transporting, ubiquitous, Calcium pump 3, EC 3.6.3, EC 3.6.3.8, sarco/endoplasmic reticulum Ca2+ ATPase, sarco/endoplasmic reticulum Ca2+ -ATPase, sarcoplasmic/endoplasmic reticulum calcium ATPase 3, SERCA3calcium-translocating P-type ATPase, SR Ca(2+)-ATPase 3
Gene Symbols ATP2A3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human SERCA3. The exact sequence is proprietary.
Molecular Weight of Antigen 109 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 489
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Chicken (88%), Rat (99%), Xenopus laevis (92%), Xenopus tropicalis (91%),Bovine (98%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.