Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERF1A Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP178392SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178392SS 0.025 mL
NBP178392 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP178392SS Supplier Novus Biologicals Supplier No. NBP178392SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERF1A Polyclonal specifically detects SERF1A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SERF1A
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:600, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:600
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. O75920
Gene Alias FAM2B, h4F5, H4F5small EDRK-rich factor 1,4F5, Protein 4F5, SERF1, SMA modifier 1, small EDRK-rich factor 1A (telomeric), SMAM1FAM2ASERF1B, spinal muscular atrophy-related gene H4F5
Gene Symbols SERF1A
Host Species Rabbit
Immunogen A synthetic peptide made to the N-terminal portion of the human SERF1A protein (between residues 1-60) [UniProt O75920]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8293
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.