Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERF1A Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
| Antigen | SERF1A |
|---|---|
| Concentration | 1.11 mg/mL |
| Dilution | Immunohistochemistry 1:600, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:600 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP178392SS
![]() |
Novus Biologicals
NBP178392SS |
0.025 mL |
Each for $207.00
|
|
|||||
NBP178392
![]() |
Novus Biologicals
NBP178392 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
SERF1A Polyclonal specifically detects SERF1A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SERF1A | |
| Immunohistochemistry 1:600, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:600 | |
| Polyclonal | |
| Rabbit | |
| Alzheimers Research, Neuroscience | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| FAM2B, h4F5, H4F5small EDRK-rich factor 1,4F5, Protein 4F5, SERF1, SMA modifier 1, small EDRK-rich factor 1A (telomeric), SMAM1FAM2ASERF1B, spinal muscular atrophy-related gene H4F5 | |
| SERF1A | |
| IgG | |
| Affinity Purified |
| 1.11 mg/mL | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| O75920 | |
| 8293 | |
| A synthetic peptide made to the N-terminal portion of the human SERF1A protein (between residues 1-60) [UniProt O75920] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title