Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERF2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP178375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178375 0.1 mL
NBP178375SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP178375 Supplier Novus Biologicals Supplier No. NBP178375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SERF2 Polyclonal specifically detects SERF2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SERF2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.16 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:300
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P84101
Gene Alias 4F5rel, FAM2CFLJ38557,4F5REL, FLJ20431, FLJ37527, Gastric cancer-related protein VRG107, H4F5REL, HsT17089, MGC48826, Protein 4F5-related, small EDRK-rich factor 2
Gene Symbols SERF2
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human SERF2 protein (between residues 1-50) [UniProt P84101]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10169
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.