Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERINC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16010220UL
Description
SERINC2 Polyclonal specifically detects SERINC2 in Human samples. It is validated for Western Blot.Specifications
SERINC2 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
FKSG84, PRO0899, serine incorporator 2, TDE2, TDE2L | |
Rabbit | |
Affinity Purified | |
RUO | |
347735 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
SERINC2 | |
Synthetic peptides corresponding to SERINC2(serine incorporator 2) The peptide sequence was selected from the N terminal of SERINC2. Peptide sequence VCEEGAGIPTVLQGHIDCGSLLGYRAVYRMCFATAAFFFFFTLLMLCVSS. | |
20 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction