Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERINC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 202661AP150UL

Description
The antibody is specific to SERINC2.
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Isoform 2 is involved in the regulation of the contraction/relaxation cycle. Wang Y. -G., Submitted (FEB-2001) to the EMBL/GenBank/DDBJ databases. Clark H. F., Genome Res. 13:2265-2270(2003). The MGC Project Team, Genome Res. 14:2121-2127(2004).Specifications
SERINC2 | |
Polyclonal | |
Unconjugated | |
SERINC2 | |
FKSG84, PRO0899, SERINC2, serine incorporator 2, TDE2, TDE2L, UNQ263/PRO300 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230779, 313057, 347735 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K0E7, Q96SA4 | |
Serinc2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction