Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERINC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SERINC2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2804
![]() ![]() |
Proteintech Group Inc
202661AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to SERINC2.
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Isoform 2 is involved in the regulation of the contraction/relaxation cycle. Wang Y. -G., Submitted (FEB-2001) to the EMBL/GenBank/DDBJ databases. Clark H. F., Genome Res. 13:2265-2270(2003). The MGC Project Team, Genome Res. 14:2121-2127(2004).Specifications
SERINC2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8K0E7, Q96SA4 | |
230779, 313057, 347735 | |
Peptide | |
Primary | |
-20°C | |
SERINC2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FKSG84, PRO0899, SERINC2, serine incorporator 2, TDE2, TDE2L, UNQ263/PRO300 | |
Serinc2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title