Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERINC3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB046338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046338 Supplier Novus Biologicals Supplier No. NBP297673JF646

Rabbit Polyclonal Antibody

SERINC3 Polyclonal specifically detects SERINC3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SERINC3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AIGP1, DIFF33tumor differentially expressed 1, placental transmembrane protein (mouse testicular tumor differentiallyexpressed), SBBI99, serine incorporator 3, TDE, TDE1Tumor differentially expressed protein 1, TMS-1, transmembrane protein SBBI99, tumour differentially expressed 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SERINC3 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10955
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.