Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serine Dehydratase Antibody (OTI3D3), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N274074PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274074PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274074PEC7 Supplier Novus Biologicals Supplier No. NBP274074PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serine Dehydratase Monoclonal specifically detects Serine Dehydratase in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Serine Dehydratase
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3D3
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 4.3.1.17, EC 4.3.1.19, L-serine ammonia-lyase, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDHL-serine dehydratase, serine dehydratase, TDH
Gene Symbols SDS
Host Species Mouse
Immunogen Full length recombinant protein of human Serine Dehydratase (NP_006834) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10993.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.