Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serine Palmitoyltransferase 2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357903 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357903 Supplier Novus Biologicals Supplier No. NBP176573MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Serine Palmitoyltransferase 2 Polyclonal antibody specifically detects Serine Palmitoyltransferase 2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Immunocytochemistry

Specifications

Antigen Serine Palmitoyltransferase 2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias EC 2.3.1, EC 2.3.1.50, KIAA0526HSN1C, LCB 2, LCB2a, LCB2NSAN1C, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, serine palmitoyltransferase 2, serine palmitoyltransferase, long chain base subunit 2, serine palmitoyltransferase, subunit II, Serine-palmitoyl-CoA transferase 2, SPT 2, SPT2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 15 amino acids near the carboxy terminus of human Serine Palmitoyltransferase 2. The immunogen is located within the last 50 amino acids of Serine Palmitoyltransferase 2. Amino Acid Sequence: LDRPFDETTYEETED
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9517
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.