Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serine racemase Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB182564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB182564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB182564 Supplier Novus Biologicals Supplier No. NBP306588JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Serine racemase Polyclonal antibody specifically detects Serine racemase in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Serine racemase
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias D-serine ammonia-lyase, D-serine dehydratase, EC 4.3.1.17, EC 4.3.1.18, EC 5.1.1.18, ILV1, ISO1, L-serine ammonia-lyase, L-serine dehydratase, serine racemase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Serine racemase fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 63826
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.