Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Antibody (YC5/45), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N106037A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106037A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106037A30 Supplier Novus Biologicals Supplier No. NB10065037AF350
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Serotonin Monoclonal specifically detects Serotonin in All species samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount.

Specifications

Antigen Serotonin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount)
Classification Monoclonal
Clone YC5/45
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 5-HT, 5-hydroxytryptamne
Host Species Rat
Immunogen Serotonin conjugated to Bovine Serum Albumin.
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-65037 recognizes serotonin and does not cross react with 5-hydroxyindolacetic acid, GABA, noradrenaline, 5-hydroxytryptophan, carnosine or melatonin. Serotonin (also known as 5-hydroxytryptamine or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotonin is widely distributed in the central and peripheral nervous system, and also in some non-neural tissues such as platelets.
Target Species All species
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.