Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Antibody (YC5/45), Biotin, Novus Biologicals™
SDP

Catalog No. NB10065037B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065037B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065037B Supplier Novus Biologicals Supplier No. NB10065037B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Serotonin Monoclonal specifically detects Serotonin in All species samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount.

Specifications

Antigen Serotonin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount)
Classification Monoclonal
Clone YC5/45
Conjugate Biotin
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Whole-Mount
Formulation PBS with 0.05% Sodium Azide
Gene Alias 5-HT, 5-hydroxytryptamne
Host Species Rat
Immunogen Serotonin conjugated to Bovine Serum Albumin.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-65037 recognizes serotonin and does not cross react with 5-hydroxyindolacetic acid, GABA, noradrenaline, 5-hydroxytryptophan, carnosine or melatonin. Serotonin (also known as 5-hydroxytryptamine or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotonin is widely distributed in the central and peripheral nervous system, and also in some non-neural tissues such as platelets.
Target Species All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.