Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Antibody (YC5/45) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10065037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065037 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065037 Supplier Novus Biologicals Supplier No. NB10065037
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Serotonin Monoclonal specifically detects Serotonin in All species, Macaca leonina (Northern Pig-Tailed Macaque), Xenopus species (African Clawed Frog) samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount.

Specifications

Antigen Serotonin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Whole-Mount)
Classification Monoclonal
Clone YC5/45
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen 1:200 - 1:500, Immunohistochemistry Whole-Mount reported in scientific literature (PMID 7781904)
Formulation PBS with 0.02% Sodium Azide
Gene Alias 5-HT, 5-hydroxytryptamne
Host Species Rat
Immunogen Serotonin conjugated to Bovine Serum Albumin.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-65037 recognizes serotonin and does not cross react with 5-hydroxyindolacetic acid, GABA, noradrenaline, 5-hydroxytryptophan, carnosine or melatonin. Serotonin (also known as 5-hydroxytryptamine or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotonin is widely distributed in the central and peripheral nervous system, and also in some non-neural tissues such as platelets.
Target Species All species, Northern Pig-Tailed Macaque (Macaca leonina), Xenopus species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.