Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serotonin Mouse anti-Human, Mouse, Porcine, Chicken, Macaque, Clone: 5HT-H209, Novus Biologicals™
SDP

Catalog No. NB141849 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB141849 500 μL
NB154851 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB141849 Supplier Novus Biologicals Supplier No. NB12016007500UL

Mouse Monoclonal Antibody has been used in 13 publications

Serotonin Monoclonal specifically detects Serotonin in Human, Mouse, Porcine, Chicken, Rhesus Macaque samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Serotonin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5HT-H209
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100
Formulation Cell culture supernatant dialysed against 0.05 mol/L Tris/HCl (pH 7.2)
Host Species Mouse
Immunogen 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin).
Purification Method Tissue culture supernatant
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Pig, Chicken, Macaque
Content And Storage Store at 4°C. Do not freeze.
Form Supernatant
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.