Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serotonin N-acetyltransferase Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303349100UL
Description
Serotonin N-acetyltransferase Polyclonal specifically detects Serotonin N-acetyltransferase in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Serotonin N-acetyltransferase | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| AA-NAT, aralkylamine N-acetyltransferaseEC 2.3.1.87, arylalkylamine N-acetyltransferase, Serotonin acetylase, SNATserotonin N-acetyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human Serotonin N-acetyltransferase (NP_001079.1). | |
| 100 μL | |
| Cancer, Circadian Rhythm, Neuroscience, Neurotransmission, Phospho Specific | |
| 15 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction