Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serotonin N-acetyltransferase Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $485.00
Specifications
| Antigen | Serotonin N-acetyltransferase |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
Serotonin N-acetyltransferase Polyclonal specifically detects Serotonin N-acetyltransferase in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Serotonin N-acetyltransferase | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AA-NAT, aralkylamine N-acetyltransferaseEC 2.3.1.87, arylalkylamine N-acetyltransferase, Serotonin acetylase, SNATserotonin N-acetyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-207 of human Serotonin N-acetyltransferase (NP_001079.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Circadian Rhythm, Neuroscience, Neurotransmission, Phospho Specific | |
| PBS with 50% glycerol, pH7.3. | |
| 15 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title