Learn More
BD Serotyping Antisera for Salmonella, Glycerinated
Quick, easy and accurate identification of Salmonella isolates
Supplier:  BD 240987
 View more versions of this product
Catalog No. 14-911-105
This item is not returnable. View return policy
Description
Description
The increased occurrence of Salmonella infections within the last decades has given rise to the need for serotyping of Salmonella as a base for proper diagnosis of such infections and their control. The Salmonella O Polyvalent antiserum is used for screening in conjunction with biochemical tests. The Salmonella Vi antiserum detects the Vi antigen, a heat-sensitive envelope antigen usually found in fresh isolates of S. typhi and S. paratyphi C.
  • Glycerinated antisera (available in vials)
  • Used in classification of Salmonella into groups A, B, C1, C2, D, E F, G, H, I, A-I & Vi and Vi according to their somatic (O) antigens
  • For in vitro diagnostic use by latex agglutination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For in vitro diagnostic use by latex agglutination
Salmonella O Group
3 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.