Learn More
BD Serotyping Antisera for Salmonella, Glycerinated
Quick, easy and accurate identification of Salmonella isolates
Specifications
Certifications/Compliance | For in vitro diagnostic use by latex agglutination |
---|---|
Product Type | Salmonella O Group |
Quantity | 3 mL |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
14-911-102
|
BD
240984 |
Salmonella O Group B; 3mL | Salmonella O Group B; 3mL | N/A | N/A | N/A | |||
14-911-103
|
BD
240985 |
Salmonella O Group C1; 3mL | Salmonella O Group C1; 3mL | N/A | N/A | N/A | |||
14-911-104
|
BD
240986 |
Salmonella O Group C2; 3mL | Salmonella O Group C2; 3mL | N/A | N/A | N/A | |||
14-911-105
|
BD
240987 |
Salmonella O Group D; 3mL | Salmonella O Group D; 3mL | N/A | N/A | N/A | |||
14-911-112
|
BD
240994 |
Salmonella O Group Vi; 3mL | Salmonella O Group Vi; 3mL | N/A | N/A | N/A | |||
Description
The increased occurrence of Salmonella infections within the last decades has given rise to the need for serotyping of Salmonella as a base for proper diagnosis of such infections and their control. The Salmonella O Polyvalent antiserum is used for screening in conjunction with biochemical tests. The Salmonella Vi antiserum detects the Vi antigen, a heat-sensitive envelope antigen usually found in fresh isolates of S. typhi and S. paratyphi C.
- Glycerinated antisera (available in vials)
- Used in classification of Salmonella into groups A, B, C1, C2, D, E F, G, H, I, A-I & Vi and Vi according to their somatic (O) antigens
- For in vitro diagnostic use by latex agglutination
Specifications
For in vitro diagnostic use by latex agglutination | |
Salmonella O Group | |
3 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.