Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A1/alpha 1-Antitrypsin Antibody (142), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB307721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307721 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307721 Supplier Novus Biologicals Supplier No. NBP289402APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Serpin A1/alpha 1-Antitrypsin Monoclonal antibody specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen Serpin A1/alpha 1-Antitrypsin
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 142
Conjugate APC
Dilution ELISA, Sandwich ELISA Capture
Formulation PBS
Gene Alias A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Serpin A1/alpha 1-Antitrypsin (Accession#: NP_000286.3; Met1-Lys418).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5265
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.