Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A1/alpha 1-Antitrypsin Antibody, Novus Biologicals™
SDP

Catalog No. NBP1780981G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1780981G 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP1780981G Supplier Novus Biologicals Supplier No. NBP1780980.1MG

Rabbit Polyclonal Antibody

Serpin A1/alpha 1-Antitrypsin Polyclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Serpin A1/alpha 1-Antitrypsin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.41 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:10000-1:600000, Immunohistochemistry 1:500-1:2500, Immunohistochemistry-Paraffin 1:500 -1:2500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P01009
Gene Alias A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Gene Symbols SERPINA1
Host Species Rabbit
Immunogen Human Alpha-1-anti-Trypsin
Purification Method Affinity Purified
Quantity 0.1 mg
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5265
Test Specificity Analysis by SDS-PAGE was used to determine that this preparation is substantially free of aggregates and shows a banding pattern consistent with purified Rabbit IgG.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.