Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Serpin A1/alpha 1-Antitrypsin Antibody (OTI9A1), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274095B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274095B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274095B Supplier Novus Biologicals Supplier No. NBP274095B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Serpin A1/alpha 1-Antitrypsin
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI9A1
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin)
Gene Symbols SERPINA1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human Serpin A1/alpha 1-Antitrypsin expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5265.0
Target Species Human, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.